N,N-Diethyl-m-toluamide ≥97%

Warning

Synonyms: Diethyl-m-toluamide, Diethyltoluamide, N,N-Diethyl-3-methylbenzamide, Deet

D100951-100G D100951-2KG D100951-500G
220120-779EA 53.95 USD
220120-779 220120-780 220120-781
N,N-Diethyl-m-toluamide ≥97%
N,N-Diethyl-m-toluamide
Formula: C₁₂H₁₇NO
MW: 191.27 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: -45 °C
Density: 0.996 g/cm³ (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR