1,12-Dodecanedioic acid ≥99%

Warning

Synonyms: Dodecanedioic acid, 1,10-Decanedicarboxylic acid

D1009-500G D1009-100G
220120-777EA 100.37 USD
220120-777 220120-776
1,12-Dodecanedioic acid ≥99%
1,12-Dodecanedioic acid
Formula: C₁₂H₂₂O₄
MW: 230.3 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 121…126 °C
Density: 1.175 g/cm³ (21 °C)
Flash Pt: 220 °C
Storage Temperature: Freezer
MDL Number: MFCD00002735
CAS Number: 693-23-2
EINECS: 211-746-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR