Diethyl sulfate ≥98%

Danger

D100706-1KG D100706-100G D100706-2KG D100706-4KG
220120-773EA 84.65 USD
220120-773 220120-772 220120-774 220120-775
Diethyl sulfate ≥98%
Diethyl sulphate
Formula: C₄H₁₀O₄S
MW: 154.19 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.179 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS Number: 64-67-5
EINECS: 200-589-6
UN: 1594
ADR: 6.1,II
Merck Index: 13,03156

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR