Cycloheptatriene

C99205-100ML C99205-1L
220117-430EA 197.08 USD
220117-430 220117-431
Cycloheptatriene
Cycloheptatriene
Formula: C₇H₈
MW: 92.14 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.8915 g/cm³ (20 °C)
Flash Pt: 3 °C
MDL Number: MFCD00004146
CAS Number: 544-25-2
EINECS: 208-866-3
UN: 2603
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR