Cycloheptanone ≥99%

Synonyms: Suberon, Cycloheptan-1-one

C99000-250G C99000-25G C99000-100G
220117-417EA 165.85 USD
220117-417 220117-418 220117-416
Cycloheptanone ≥99%
Cycloheptanone
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 179 °C (1013 hPa)
Melting Pt: –21 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00004159
CAS Number: 502-42-1
EINECS: 207-937-6
UN: 1224
ADR: 3,III
Merck Index: 13,02751

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR