Colchicine

Danger

Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine

C9754-5G C9754-100MG C9754-1G C9754-500MG
220117-354EA 571.52 USD
220117-354 220117-351 220117-352 220117-353
Colchicine
Colchicine
Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
Melting Pt: 154 - 156 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078484
CAS Number: 64-86-8
EINECS: 200-598-5
UN: 1544
ADR: 6.1,I
Merck Index: 12,02536

Order Now

Specification Test Results

Assay 95% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR