Oil of castor

Synonyms: Castor oil, Ricinus oil, Oleum Ricini s. Castoris, Oil of Palma Christi

C9606-500ML C9606-1L
220115-997EA 168.62 USD
220115-997 220115-996
Oil of castor
Oil of castor
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: >113 °C (closed cup)
MDL Number: MFCD00130746
CAS Number: 8001-79-4
EINECS: 232-293-8
Merck Index: 13,01908

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR