Cyclobutanone ≥99%

C96001-1G C96001-5G
220117-304EA 34.57 USD
220117-304 220117-305
Cyclobutanone ≥99%
Cyclobutanone
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 99 °C (1013 hPa)
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001332
CAS Number: 1191-95-3
EINECS: 214-745-6
UN: 1224
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR