Cyclobutanecarboxylic acid ≥98%

Danger

C95609-25G C95609-5G C95609-100G
220117-292EA 75.71 USD
220117-292 220117-293 220117-291
Cyclobutanecarboxylic acid ≥98%
Cyclobutanecarboxylic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: –20 °C
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001323
CAS Number: 3721-95-7
EINECS: 223-072-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR