4-Hydroxybenzonitrile ≥95%

Warning

Synonyms: p-Cyanophenol, 4-Cyanophenol, p-Hydroxybenzonitrile

C94009-5G C94009-100G C94009-25G
220117-256EA 37.69 USD
220117-256 220117-254 220117-255
4-Hydroxybenzonitrile ≥95%
4-Hydroxybenzonitrile
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 148 °C (1 mmHg)
Melting Pt: 109…112 °C
Density: ~ 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR