Indole-5-carbonitrile

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

C92006-5G C92006-10G
220117-163EA 74.09 USD
220117-163 220117-162
Indole-5-carbonitrile
Indole-5-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 106…108 °C
MDL Number: MFCD00005669
CAS Number: 15861-24-2
EINECS: 239-986-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR