4-Chloro-1-naphthol

Synonyms: 4-Chloronaphthalen-1-ol, 1-Chloro-4-hydroxynaphthalene, 4-CN, Chloronaphthol

C8890-100G C8890-2.5G C8890-25G C8890-5G
220116-823EA 1569.23 USD
220116-823 220116-824 220116-825 220116-826
4-Chloro-1-naphthol
4-Chloro-1-naphthol
Formula: ClC₁₀H₆OH
MW: 178.62 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 123 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003974
CAS Number: 604-44-4
EINECS: 210-068-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR