Cyanoacetic acid ≥99%

Danger

C88505-1KG C88505-25G C88505-500G
220117-059EA 232.62 USD
220117-059 220117-060 220117-061
Cyanoacetic acid ≥99%
Cyanoacetic acid
Formula: C₃H₃NO₂
MW: 85.06 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 66 °C
Density: Min. 1.1 g/cm³ (20 °C)
Flash Pt: >165 °C
MDL Number: MFCD00002677
CAS Number: 372-09-8
EINECS: 206-743-9
UN: 3261
ADR: 8,II
Merck Index: 13,02718

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR