Copper(II) acetylacetonate

Warning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

C87851-500G C87851-100G C87851-25G
220117-043EA 484.62 USD
220117-043 220117-041 220117-042
Copper(II) acetylacetonate
Copper(II) acetylacetonate
Formula: C₁₀H₁₄CuO₄
MW: 261.76 g/mol
Melting Pt: 279…283 °C
MDL Number: MFCD00000016
CAS Number: 13395-16-9
EINECS: 236-477-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR