Cesium hydroxide hydrate ≥90%

C8518-10G C8518-50G C8518-250G
220114-786EA 72.31 USD
220114-786 220114-788 220114-787
Cesium hydroxide hydrate ≥90%
Cesium hydroxide hydrate
Formula: HCsO·H₂O
MDL Number: MFCD00010964
CAS Number: 12260-45-6
EINECS: 244-344-1
UN: 2682
ADR: 8,II

Order Now

Specification Test Results

Assay 90%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR