Citraconic acid ≥98%

Synonyms: 2-Methyl-2-butenedioic acid

C82604-100G C82604-5G
220115-722EA 185.23 USD
220115-722 220115-723
Citraconic acid ≥98%
Citraconic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 98 - 99 °C
Density: 1.617 g/cm³ (20 °C)
MDL Number: MFCD00078086
CAS Number: 498-23-7
EINECS: 207-858-7
Merck Index: 13,02345

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR