trans-Cinnamic acid

Warning

Synonyms: Cinnamic acid (only trans), (2E)-2-Phenyl-2-propenoic acid, trans-Cinnamic acid

C80857-5G C80857-250G
220116-742EA 47.6 USD
220116-742 220116-106
trans-Cinnamic acid
trans-Cinnamic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 131…134 °C
Density: 1.25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR