Cholesterol

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

C75209-25G C75209-500G
220116-541EA 447.23 USD
220116-541 220116-540 220116-542
Cholesterol
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 147…150 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR