α-Chloro-p-xylene ≥98%

Warning

Synonyms: alpha-Chloro-p-xylene, 4-Methylbenzyl chloride

C73400-5G C73400-100G
220116-480EA 102.74 USD
220116-480 220116-479
α-Chloro-p-xylene ≥98%
α-Chloro-p-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000919
CAS Number: 104-82-5
EINECS: 203-241-1
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR