α-Chloro-m-xylene ≥98%

Warning

Synonyms: 1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride

C73354-100G C73354-25G
220116-477EA 216.46 USD
220116-477 220116-478
α-Chloro-m-xylene ≥98%
α-Chloro-m-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 196 °C (1013 hPa)
Density: 1.064 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR