4-Chloroquinoline ≥99%

Warning

C70509-5G C70509-1G
220116-376EA 189.54 USD
220116-376 220116-375
4-Chloroquinoline ≥99%
4-Chloroquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 260 - 261 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006773
CAS Number: 611-35-8
EINECS: 210-267-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR