3-Chloropyridine

C70002-25G C70002-50G
220116-348EA 68.6 USD
220116-348 220116-349
3-Chloropyridine
3-Chloropyridine
Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 148 °C (1013 hPa)
Melting Pt: -0.5 °C
Density: 1.194 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006375
CAS Number: 626-60-8
EINECS: 210-955-7
UN: 1100
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR