4'-Chloropropiophenone ≥98%

Synonyms: 1-(4-Chlorophenyl)propan-1-one

C69209-100G C69209-25G
220116-324EA 143.08 USD
220116-324 220116-325
4'-Chloropropiophenone ≥98%
4'-Chloropropiophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 277 °C (1013 hPa)
Melting Pt: 33 - 34 °C
MDL Number: MFCD00000626
CAS Number: 6285-05-8
EINECS: 228-511-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR