2-Chloropropane ≥99%

Danger

C68563-500ML C68563-1L
220116-299EA 920 USD
220116-299 220116-298
2-Chloropropane ≥99%
2-Chloropropane
Formula: C₃H₇Cl
MW: 78.54 g/mol
Boiling Pt: 35.74 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: –42 °C
Storage Temperature: Ambient
MDL Number: MFCD00000867
CAS Number: 75-29-6
EINECS: 200-858-8
UN: 2356
ADR: 3,I
Merck Index: 13,05231

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR