(3-Chloropropyl)benzene ≥99%

Synonyms: 1-Chloro-3-phenylpropane, 3-Phenylpropyl chloride, 3-Phenylpropyl Chloride

C68105-5ML C68105-100ML
220116-282EA 34.35 USD
220116-282 220116-281
(3-Chloropropyl)benzene ≥99%
(3-Chloropropyl)benzene
Formula: ClH₂CCH₂CH₂(C₆H₅)
MW: 154.64 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001001
CAS Number: 104-52-9
EINECS: 203-210-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR