5-Chloro-2-pentanone ≥85%, Technical Grade

Warning

Synonyms: 5-Chloropentan-2-one

C62603-25G C62603-100G
220116-060EA 53.95 USD
220116-060 220116-059
5-Chloro-2-pentanone ≥85%, Technical Grade
5-Chloro-2-pentanone
Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 71 °C (1013 hPa)
Melting Pt: -48 °C
Density: 1.057 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001008
CAS Number: 5891-21-4
EINECS: 227-565-8
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR