1-Chloro-3-pentanone ≥85%, Technical Grade

C62506-25G C62506-5G
220116-056EA 495.62 USD
220116-056 220116-057
1-Chloro-3-pentanone ≥85%, Technical Grade
1-Chloro-3-pentanone
Formula: C₅H₉ClO
MW: 120.58 g/mol
Storage Temperature: Refrigerator
CAS Number: 32830-97-0
EINECS: 251-247-8
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR