2-Chloro-5-nitropyridine

Warning

C61801-5G C61801-25G
220116-044EA 63.65 USD
220116-044 220116-043
2-Chloro-5-nitropyridine
2-Chloro-5-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR