Crystal violet ACS

Danger

Synonyms: CI 42555

C6158-100G C6158-50G
220116-029EA 190.4 USD
220116-029 220116-030
Crystal violet ACS
Crystal violet
Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 189…194 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9d5cb226f527b08e891a392ec4cc3516751db1c&vid=8c64844955baf8e8401738ed1a85dc2ad26f8921&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR