2-Chloro-4-nitrophenol ≥97%

Synonyms: 1-Chloro-2-hydroxy-5-nitrobenzene

C61208-10G C61208-50G
220115-970EA 37.15 USD
220115-970 220115-971
2-Chloro-4-nitrophenol ≥97%
2-Chloro-4-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 109…112 °C
MDL Number: MFCD00043910
CAS Number: 619-08-9
EINECS: 210-578-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR