5-Chloro-2-nitroanisole ≥97%

C58606-10G C58606-25G
220115-554EA 97.03 USD
220115-554 220115-555
5-Chloro-2-nitroanisole ≥97%
5-Chloro-2-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 71 °C
MDL Number: MFCD00007288
CAS Number: 6627-53-8
EINECS: 229-594-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR