2-Chloro-2-methylpropane ≥99%

Danger

Synonyms: tert-Butyl chloride

C56352-25ML C56352-2L C56352-500ML
220115-749EA 33.6 USD
220115-749 220115-750 220115-751
2-Chloro-2-methylpropane ≥99%
2-Chloro-2-methylpropane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 51 °C (1013 hPa)
Melting Pt: -28 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: -27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000816
CAS Number: 507-20-0
EINECS: 208-066-4
UN: 1127
ADR: 3,II
Merck Index: 13,01561

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR