4-Chloro-m-cresol ≥99%

Danger

Synonyms: 4-Chloro-3-methylphenol, 2-Chloro-5-hydroxytoluene, 1-Chloro-4-hydroxy-2-methylbenzene, p-Chloro m-cresol

C55402-500G C55402-5G C55402-100G
220115-205EA 85.51 USD
220115-205 220115-206 220115-203
4-Chloro-m-cresol ≥99%
4-Chloro-m-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 63 - 65 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3437
ADR: 6.1,II
Merck Index: 13,02152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR