tri-Calcium phosphate

C5267-100G C5267-500G
220115-535EA 178.46 USD
220115-535 220115-536
tri-Calcium phosphate
tri-Calcium phosphate
Formula: Ca₃(PO₄)₂
Density: 3.14 g/cm³ (20 °C)
MDL Number: MFCD00015984
CAS Number: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR