Sodium octanoate

Warning

Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate

C5038-100G C5038-500G C5038-10G C5038-1KG
220115-449EA 124.78 USD
220115-449 220115-452 220115-450 220115-451
Sodium octanoate
Sodium octanoate
Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
Melting Pt: 225 - 355 °C
Density: Min. 0.9949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR