4-Chloro-4'-fluorobutyrophenone ≥97%

Synonyms: 4-Chloro-1-(4-fluorophenyl)butan-1-one, ω-Chloro-4-fluorobutyrophenone, γ-Chloro-4-fluorobutyrophenone

C43609-25G C43609-100G
220113-649EA 56.22 USD
220113-649 220113-648
4-Chloro-4'-fluorobutyrophenone ≥97%
4-Chloro-4'-fluorobutyrophenone
Formula: C₁₀H₁₀ClFO
MW: 200.64 g/mol
Boiling Pt: 300 °C (1013 hPa)
Density: 1.2267 g/cm³ (20 °C)
MDL Number: MFCD00001007
CAS Number: 3874-54-2
EINECS: 223-395-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR