Coumarin

Warning

C4261-50G C4261-100G C4261-500G
220115-123EA 64.03 USD
220115-123 220115-121 220115-122
Coumarin
Coumarin
Formula: C₉H₆O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 67…69 °C
Density: 0.935 g/cm³ (20 °C)
Flash Pt: 162 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR