2-Chloroethyl ethyl ether ≥99%

Synonyms: 2-Ethoxyethyl chloride, 2-Chlorodiethyl ether, 2-Chloroethylethylether

C41207-25G C41207-100G
220111-458EA 1415.38 USD
220111-458 220111-457
2-Chloroethyl ethyl ether ≥99%
2-Chloroethyl ethyl ether
Formula: C₄H₉ClO
MW: 108.57 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: –60 °C
Density: 1.0572 g/cm³ (0 °C)
MDL Number: MFCD00000974
CAS Number: 628-34-2
EINECS: 211-037-9
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR