Bis(2-chloroethyl)ether ≥99%

Synonyms: 2-Chloroethyl ether

C41134-500G C41134-100G
220115-059EA 146.46 USD
220115-059 220115-058
Bis(2-chloroethyl)ether ≥99%
Bis(2-chloroethyl)ether
Formula: C₄H₈Cl₂O
MW: 143.01 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1,II
Merck Index: 13,03091

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR