Calcium sulfate dihydrate ≥99%

Synonyms: Plaster of Paris

C3771-500G C3771-1KG
220114-928EA 42.22 USD
220114-928 220114-927
Calcium sulfate dihydrate ≥99%
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR