Cytosine

Warning

C3506-100G C3506-1G C3506-25G C3506-5G C3506-10G
220114-776EA 1280.77 USD
220114-776 220114-819 220114-820 220114-821 220114-818
Cytosine
Cytosine
Formula: C₄H₅N₃O
Melting Pt: 313 - 316 °C
Density: 0.99771 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006034
CAS Number: 71-30-7
EINECS: 200-749-5
Merck Index: 13,02824

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR