Chlorodifluoroacetic acid ≥98%

Danger

C34200-25G C34200-100G
220113-883EA 46.03 USD
220113-883 220113-882
Chlorodifluoroacetic acid ≥98%
Chlorodifluoroacetic acid
Formula: C₂HClF₂O₂
Boiling Pt: 122 °C (1013 hPa)
Density: 2.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004176
CAS Number: 76-04-0
EINECS: 200-928-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR