2-Chlorocyclohexanone ≥98%

C32607-25G C32607-100G C32607-5G
220114-730EA 263.85 USD
220114-730 220114-729 220114-731
2-Chlorocyclohexanone ≥98%
2-Chlorocyclohexanone
Formula: C₆H₉ClO
MW: 132.59 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 18…20 °C
Density: 1.16 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001626
CAS Number: 822-87-7
EINECS: 212-505-5
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR