3-Chloro-2-(chloromethyl)propene ≥99%

Synonyms: 3-Chloro-2-chloromethyl-1-propene, 3-Chloro-2-(chloromethyl)-1-propene

C31104-50G C31104-10G
220114-381EA 538.46 USD
220114-381 220114-380
3-Chloro-2-(chloromethyl)propene ≥99%
3-Chloro-2-(chloromethyl)propene
Formula: C₄H₆Cl₂
MW: 125 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: -11 °C
Density: 1.175 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000952
CAS Number: 1871-57-4
EINECS: 217-489-3
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR