2-Chlorobutane ≥99%

C28898-100ML C28898-250ML
220114-589EA 32.09 USD
220114-589 220114-590
2-Chlorobutane ≥99%
2-Chlorobutane
Formula: C₄H₉Cl
MW: 92.57 g/mol
Boiling Pt: 68 °C (1013 hPa)
Melting Pt: –132 °C
Density: 0.87 g/cm³ (20 °C)
Flash Pt: –10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000871
CAS Number: 78-86-4
EINECS: 201-151-7
UN: 1127
ADR: 3,II
Merck Index: 13,01560

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR