Caprylic acid ≥99%

Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

C2875-100ML C2875-500ML C2875-1L C2875-10ML
220114-574EA 141.54 USD
220114-574 220114-577 220114-576 220114-575
Caprylic acid ≥99%
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR