Calcium nitrate tetrahydrate

Warning

C2786-1KG C2786-500G
220114-537EA 171.22 USD
220114-537 220114-538
Calcium nitrate tetrahydrate
Calcium nitrate tetrahydrate
Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Melting Pt: ~ 42 °C
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 13,01687

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR