4-Chlorobenzyl alcohol ≥99%

Synonyms: (4-Chlorophenyl)methanol

C27115-100G C27115-25G
220114-174EA 453.38 USD
220114-174 220114-175
4-Chlorobenzyl alcohol ≥99%
4-Chlorobenzyl alcohol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 234 °C (1013 hPa)
Melting Pt: 67…70 °C
MDL Number: MFCD00004652
CAS Number: 873-76-7
EINECS: 212-852-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR