4-Chlorobenzophenone ≥99%

Synonyms: p-Chlorobenzophenone

C25007-100G C25007-5G
220114-358EA 61.92 USD
220114-358 220114-359
4-Chlorobenzophenone ≥99%
4-Chlorobenzophenone
Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00000622
CAS Number: 134-85-0
EINECS: 205-160-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR