3-Chlorobenzoic acid ≥99%

Warning

C24604-25G C24604-100G
220114-335EA 43.08 USD
220114-335 220114-334
3-Chlorobenzoic acid ≥99%
3-Chlorobenzoic acid
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 122 °C
Density: 1.496 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002491
CAS Number: 535-80-8
EINECS: 208-618-4
Merck Index: 13,02142

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR